This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03931104_s1
See other C030015E24RIK GE Assays ›
Gene Symbol:
C030015E24Rik
Gene Name:
RIKEN cDNA C030015E24 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03931104_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: C030015E24Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK021085.1 - 1 55 94
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY728168.1 - 1 52 Chr. 5 - 48997851 - 48998536 [-] on Build GRCm38 94

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream Kv channel interacting protein 4 Kcnip4 AV032399,
Calp,
Calp250,
KChIP4,
KChIP4a
Sense Chr. 5 - 48389503 - 49525122 on NCBI Build GRCm38
Downstream adhesion G protein-coupled receptor A2 Adgra2 3830613O22Rik,
AU044632,
Gpr125,
Tem5-like
961.4kb Sense Chr. 5 - 49959951 - 50058996 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Illumina

Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 100580068 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0