This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03931075_s1
See other C030014A21RIK GE Assays ›
Gene Symbol:
C030014A21Rik
Gene Name:
RIKEN cDNA C030014A21 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03931075_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: C030014A21Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK021077.1 - 1 250 101
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY728157.1 - 1 247 Chr. 13 - 117822316 - 117822978 [+] on Build GRCm38 101

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream hyperpolarization-activated,
cyclic nucleotide-gated K+ 1
Hcn1 Bcng1,
C630013B14Rik,
HAC2
Sense Chr. 13 - 117602320 - 117987414 on NCBI Build GRCm38
Downstream mitochondrial ribosomal protein S30 Mrps30 2610020A16Rik,
AA968347,
PAP,
Pdcd9
556.8kb Anti-sense Chr. 13 - 118380110 - 118387252 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix ABI

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2194054 AK021077.1 mouse_transcriptome
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1453897_PM_at - -
Mouse Genome 430 2.0 Array 1453897_PM_at - -
Mouse Genome 430 2.0 Array 1453897_at - -
Mouse Expression Set 430 1453897_PM_at - -
Mouse Expression Set 430 1453897_at - -
Affymetrix HT Mouse Genome 430B Array 1453897_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1453897_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 477945 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0