This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03931062_s1
See other C030013C21RIK GE Assays ›
Gene Symbol:
C030013C21Rik
Gene Name:
RIKEN cDNA C030013C21 gene
Gene Aliases:
-
Chromosome Location:
Chr.11: 80507410 - 80509106 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03931062_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: C030013C21Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK021072.1 - 1 250 78
AV253635.2 - 1 331 78
BB614470.1 - 1 470 78
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY728151.1 - 1 249 Chr. 11 - 80508483 - 80508885 [-] on Build GRCm38 78
GenBankmRNA BB356264.1 - 1 156 Chr. 11 - 80508498 - 80508792 [-] on Build GRCm38 78

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream cyclin-dependent kinase 5,
regulatory subunit 1 (p35)
Cdk5r1 Cdk5r,
D11Bwg0379e,
p25,
p35
-27.3kb Anti-sense Chr. 11 - 80477046 - 80481179 on NCBI Build GRCm38
Downstream transmembrane protein 98 Tmem98 6530411B15Rik,
AI463522
301.5kb Anti-sense Chr. 11 - 80810415 - 80822033 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2317332 AV253635.2 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2317332 AV253635.2 mouse_transcriptome
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1444183_PM_at - -
Mouse Genome 430 2.0 Array 1444183_PM_at - -
Mouse Genome 430 2.0 Array 1444183_at - -
Mouse Expression Set 430 1444183_at - -
Mouse Expression Set 430 1444183_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1444183_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1444183_at - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0