This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03931026_s1
See other C030010C08RIK GE Assays ›
Gene Symbol:
C030010C08Rik
Gene Name:
RIKEN cDNA C030010C08 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03931026_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: C030010C08Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK021060.1 - 446 82
AK135037.1 - 894 82
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY728129.1 - 1 442 Chr. 13 - 63814454 - 63815110 [-] on Build GRCm38 82

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream excision repair cross-complementing rodent repair deficiency,
complementation group 6 like 2
Ercc6l2 0610007P08Rik,
1700019D06Rik,
9330134C04Rik,
Rad26l,
Sr278
Anti-sense Chr. 13 - 63813415 - 63900302 on NCBI Build GRCm38
Downstream hydroxysteroid (17-beta) dehydrogenase 3 Hsd17b3 243.2kb Sense Chr. 13 - 64058274 - 64089258 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix ABI

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1975355 AK135037.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 687726 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1975355 AK135037.1 mouse_transcriptome
Agilent Six_ChIP_03_of_03_v2 A_68_P29095464 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29095460 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29095472 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29095468 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29095465 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29095469 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29095463 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29095467 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29095461 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29095474 - -
Array Name Probe IDs Targets Target Type
Murine Genome U74v2 Set 137187_PM_at - -
Murine Genome U74v2 Set 137187_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 856039 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0