This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03930986_s1
See other C030004M13RIK GE Assays ›
Gene Symbol:
C030004M13Rik
Gene Name:
RIKEN cDNA C030004M13 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03930986_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: C030004M13Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK021045.1 - 1 754 73

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream phosphodiesterase 10A Pde10a Sense Chr. 17 - 8525187 - 8986648 on NCBI Build GRCm38
Downstream RIKEN cDNA 1700010I14 gene 1700010I14Rik 50.0kb Sense Chr. 17 - 8988320 - 9008319 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1433332_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1433333_PM_at - -
Mouse Genome 430 2.0 Array 1433332_PM_at - -
Mouse Genome 430 2.0 Array 1433333_at - -
Mouse Genome 430 2.0 Array 1433332_at - -
Mouse Genome 430 2.0 Array 1433333_PM_at - -
Mouse Expression Set 430 1433332_at - -
Mouse Expression Set 430 1433333_at - -
Mouse Expression Set 430 1433333_PM_at - -
Mouse Expression Set 430 1433332_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1433332_at - -
Affymetrix HT Mouse Genome 430B Array 1433332_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1433333_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1433333_at - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 103130463 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0