This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03930876_s1
See other B230112I24RIK GE Assays ›
Gene Symbol:
B230112I24Rik
Gene Name:
RIKEN cDNA B230112I24 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03930876_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: B230112I24Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK020969.1 - 1 556 92
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY727025.1 - 1 556 Chr. 6 - 45073566 - 45074215 [+] on Build GRCm38 92

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream thiamine pyrophosphokinase Tpk1 -1.4mb Anti-sense Chr. 6 - 43344999 - 43666484 on NCBI Build GRCm38
Downstream cullin 1 Cul1 2.4mb Sense Chr. 6 - 47452273 - 47526139 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1433187_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1433188_PM_at - -
Mouse Genome 430 2.0 Array 1433188_PM_at - -
Mouse Genome 430 2.0 Array 1433187_at - -
Mouse Genome 430 2.0 Array 1433188_at - -
Mouse Genome 430 2.0 Array 1433187_PM_at - -
Mouse Expression Set 430 1433188_at - -
Mouse Expression Set 430 1433187_PM_at - -
Mouse Expression Set 430 1433188_PM_at - -
Mouse Expression Set 430 1433187_at - -
Affymetrix HT Mouse Genome 430B Array 1433188_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1433187_at - -
Affymetrix HT Mouse Genome 430B Array 1433187_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1433188_at - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 102970736 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0