This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03930769_s1
See other A930036A04RIK GE Assays ›
Gene Symbol:
A930036A04Rik
Gene Name:
RIKEN cDNA A930036A04 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03930769_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: A930036A04Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK020933.1 - 1 652 99

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream legumain Lgmn AEP,
AI746452,
AU022324,
Prsc1
-1.0kb Anti-sense Chr. 12 - 102394098 - 102439697 on NCBI Build GRCm38
Downstream golgi autoantigen,
golgin subfamily a,
5
Golga5 Ret-II 27.4kb Sense Chr. 12 - 102469134 - 102497911 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 292723 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28691556 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28691559 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28691555 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28691562 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10382528 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1454571_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1433347_PM_at - -
Mouse Genome 430 2.0 Array 1454571_PM_at - -
Mouse Genome 430 2.0 Array 1454571_at - -
Mouse Genome 430 2.0 Array 1433347_PM_at - -
Mouse Genome 430 2.0 Array 1433347_at - -
Mouse Expression Set 430 1454571_PM_at - -
Mouse Expression Set 430 1433347_PM_at - -
Mouse Expression Set 430 1454571_at - -
Mouse Expression Set 430 1433347_at - -
Affymetrix HT Mouse Genome 430B Array 1454571_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1433347_at - -
Affymetrix HT Mouse Genome 430B Array 1454571_at - -
Affymetrix HT Mouse Genome 430B Array 1433347_PM_at - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 105340079 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0