This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03930756_s1
See other A930033M24RIK GE Assays ›
Gene Symbol:
A930033M24Rik
Gene Name:
RIKEN cDNA A930033M24 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03930756_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: A930033M24Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK020926.1 - 1 155 97
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY742326.1 - 1 155 Chr. 6 - 40470590 - 40471280 [-] on Build GRCm38 97

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream WEE1 homolog 2 (S. pombe) Wee2 Gm1065,
Wee1B
-3.7kb Anti-sense Chr. 6 - 40426699 - 40466815 on NCBI Build GRCm38
Downstream single-stranded DNA binding protein 1 Ssbp1 2810480P10Rik,
G630031O20Rik,
MtSSB,
mtDBP
135.0 Anti-sense Chr. 6 - 40471415 - 40481825 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Agilent Illumina

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 503881 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 123950 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05873891 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 106380332 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0