This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03930755_s1
See other A930033M14RIK GE Assays ›
Gene Symbol:
A930033M14Rik
Gene Name:
RIKEN cDNA A930033M14 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03930755_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: A930033M14Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AA547551.1 - 1 125 83
AI504129.1 - 1 132 83
AK020925.1 - 1 151 83
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB284009.1 - 1 162 Chr. 10 - 40260945 - 40261333 [+] on Build GRCm38 83
GenBankmRNA BY742325.1 - 1 151 Chr. 10 - 40260934 - 40261333 [+] on Build GRCm38 83

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream U3A small nuclear RNA Rnu3a -2.3kb Sense Chr. 10 - 40258576 - 40258679 on NCBI Build GRCm38
Downstream S-adenosylmethionine decarboxylase 1 Amd1 1,
AdoMetDC,
Amd-1,
SAMDC,
SAMDC 1,
adoMetDC1
26.1kb Anti-sense Chr. 10 - 40287458 - 40302188 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse cDNA Microarray (G4104A) 961272 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27083158 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27083152 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27083157 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08541675 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08471556 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27083154 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27083150 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08455340 - -
Array Name Probe IDs Targets Target Type
Murine Genome U74v2 Set 135587_at - -
Murine Genome U74v2 Set 135587_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 524843 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 101090037 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0