This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03930623_s1
See other A930008B05RIK GE Assays ›
Gene Symbol:
A930008B05Rik
Gene Name:
RIKEN cDNA A930008B05 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03930623_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: A930008B05Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK020828.1 - 1 945 113

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream tumor necrosis factor receptor superfamily,
member 21
Tnfrsf21 AA959878,
DR6,
R74815,
TR7
-42.9kb Anti-sense Chr. 17 - 43016555 - 43089188 on NCBI Build GRCm38
Downstream RIKEN cDNA E130008D07 gene E130008D07Rik 12.9kb Sense Chr. 17 - 43146041 - 43158291 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix ABI

Array Name Probe IDs Targets Target Type
Agilent Six_ChIP_03_of_03_v2 A_68_P31187813 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31187808 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31187829 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13550068 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13106259 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31187810 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31187833 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31187826 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13550073 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31187831 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31187806 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13139988 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1431494_PM_at - -
Mouse Genome 430 2.0 Array 1431494_PM_at - -
Mouse Genome 430 2.0 Array 1431494_at - -
Mouse Expression Set 430 1431494_PM_at - -
Mouse Expression Set 430 1431494_at - -
Affymetrix HT Mouse Genome 430B Array 1431494_at - -
Affymetrix HT Mouse Genome 430B Array 1431494_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 807706 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0