This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03930539_s1
See other 2310029O18RIK GE Assays ›
Gene Symbol:
2310029O18Rik
Gene Name:
RIKEN cDNA 2310029O18 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03930539_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 2310029O18Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK009532.1 - 1 1943 110

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream nucleolar protein family 6 (RNA-associated) Nol6 AA410091,
Nrap
-53.1kb Anti-sense Chr. 4 - 41114427 - 41124339 on NCBI Build GRCm38
Downstream ubiquitin-associated protein 2 Ubap2 1190005K07Rik,
AA408600,
AU045235,
UBAP-2,
mKIAA1491
14.7kb Anti-sense Chr. 4 - 41194313 - 41275321 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix

Array Name Probe IDs Targets Target Type
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109166 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1454031_PM_at - -
Mouse Genome 430 2.0 Array 1454031_PM_at - -
Mouse Genome 430 2.0 Array 1454031_at - -
Mouse Expression Set 430 1454031_PM_at - -
Mouse Expression Set 430 1454031_at - -
Murine Genome U74v2 Set 170256_at - -
Murine Genome U74v2 Set 170256_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1454031_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1454031_at - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0