This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03930417_s1
See other TRAF6 GE Assays ›
Gene Symbol:
Traf6
Gene Name:
TNF receptor-associated factor 6
Gene Aliases:
2310003F17Rik, AI851288, C630032O20Rik
Chromosome Location:
Chr.2: 101678420 - 101701668 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03930417_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Traf6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK009110.1 - 1 143 59
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY708950.1 - 1 143 Chr. 2 - 101678465 - 101679125 [+] on Build GRCm38 59

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream recombination activating gene 1 Rag1 Rag-1 -28.9kb Anti-sense Chr. 2 - 101638252 - 101649532 on NCBI Build GRCm38
Downstream proline rich 5 like Prr5l 2600010E01Rik,
4833411O04Rik,
Protor-2,
Protor2
35.2kb Anti-sense Chr. 2 - 101714280 - 101883074 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7bff9b55f5-ncxmj:80/100.66.128.142:80.
git-commit: 5f36da1a8ad9bc9722227bfd86ebc827ae27d083
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0