This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03930315_s1
See other 2210411G17RIK GE Assays ›
Gene Symbol:
2210411G17Rik
Gene Name:
RIKEN cDNA 2210411G17 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03930315_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 2210411G17Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK008898.1 - 1 1052 91

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream solute carrier family 12,
member 2
Slc12a2 9330166H04Rik,
Nkcc1,
mBSC2,
sy-ns
-3.1kb Sense Chr. 18 - 57878513 - 57946821 on NCBI Build GRCm38
Downstream fibrillin 2 Fbn2 BC063774,
Fib-2,
mKIAA4226,
sne,
sy
56.5kb Anti-sense Chr. 18 - 58008623 - 58209926 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix ABI

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 384247 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 572889 - -
Whole Mouse Genome G4122A A_52_P1002658 AK008898.1 mouse_transcriptome
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1432667_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1432668_PM_at - -
Mouse Genome 430 2.0 Array 1432667_PM_at - -
Mouse Genome 430 2.0 Array 1432668_PM_at - -
Mouse Genome 430 2.0 Array 1432668_at - -
Mouse Genome 430 2.0 Array 1432667_at - -
Mouse Expression Set 430 1432667_at - -
Mouse Expression Set 430 1432667_PM_at - -
Mouse Expression Set 430 1432668_at - -
Mouse Expression Set 430 1432668_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1432667_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1432668_at - -
Affymetrix HT Mouse Genome 430B Array 1432668_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1432667_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 377946 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0