This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03930180_s1
See other 2010110E17RIK GE Assays ›
Gene Symbol:
2010110E17Rik
Gene Name:
RIKEN cDNA 2010110E17 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03930180_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 2010110E17Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK008378.1 - 1 644 75
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY708292.1 - 1 643 Chr. 8 - 86885688 - 86886541 [+] on Build GRCm38 75

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream NEDD4 binding protein 1 N4bp1 AI481586,
C81621
-430.0 Anti-sense Chr. 8 - 86841136 - 86885258 on NCBI Build GRCm38
Downstream predicted gene 5118 Gm5118 A330085E11,
EG330819
206.7kb Sense Chr. 8 - 87093310 - 87101571 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Agilent Illumina

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 651830 AK041412.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 651830 NM_177378.3 refseq_mrna_mouse_NM
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 159357 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01787279 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26031876 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07021479 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26031852 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26031858 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26031871 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07320462 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06817931 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26031888 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26031855 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26031878 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07021486 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26031880 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26031873 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26031875 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26031857 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26031864 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26031870 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26031869 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26031868 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26031860 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26031863 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26031859 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26031885 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26031887 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26031853 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 104540273 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0