This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03930100_s1
See other 2010003K10RIK GE Assays ›
Gene Symbol:
2010003K10Rik
Gene Name:
RIKEN cDNA 2010003K10 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03930100_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 2010003K10Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK008071.1 - 1 957 108

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream UDP-Gal:betaGlcNAc beta 1,4-galactosyltransferase,
polypeptide 5
B4galt5 9430078I07Rik,
AW049941,
AW539721
Sense Chr. 2 - 167298444 - 167349178 on NCBI Build GRCm38
Downstream solute carrier family 9 (sodium/hydrogen exchanger),
member 8
Slc9a8 1200006P13Rik,
6430709P13Rik,
AI182282,
NHE-8,
NHE8
95.9kb Anti-sense Chr. 2 - 167420056 - 167477000 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1431563_PM_at - -
Mouse Genome 430 2.0 Array 1431563_at - -
Mouse Genome 430 2.0 Array 1431563_PM_at - -
Mouse Expression Set 430 1431563_PM_at - -
Mouse Expression Set 430 1431563_at - -
Murine Genome U74v2 Set 166952_PM_i_at - -
Murine Genome U74v2 Set 166952_i_at - -
Murine Genome U74v2 Set 166953_PM_r_at - -
Murine Genome U74v2 Set 166953_r_at - -
Affymetrix HT Mouse Genome 430B Array 1431563_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1431563_at - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0