This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03929976_s1
See other 1810028F09RIK GE Assays ›
Gene Symbol:
1810028F09Rik
Gene Name:
RIKEN cDNA 1810028F09 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03929976_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 1810028F09Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK007631.1 - 1 147 94

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream ribonuclease,
RNase A family,
6
Rnase6 9530043P15Rik,
BB119466
-10.9kb Anti-sense Chr. 14 - 51123913 - 51132187 on NCBI Build GRCm38
Downstream ribonuclease,
RNase A family,
1 (pancreatic)
Rnase1 AI574248,
Rib-1,
Rib1
883.0 Sense Chr. 14 - 51145002 - 51146767 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix ABI

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 318085 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 78966 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08110202 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1430027_PM_at - -
Mouse Genome 430 2.0 Array 1430027_PM_at - -
Mouse Genome 430 2.0 Array 1430027_at - -
Mouse Expression Set 430 1430027_PM_at - -
Mouse Expression Set 430 1430027_at - -
Murine Genome U74v2 Set 167137_at - -
Murine Genome U74v2 Set 167137_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1430027_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1430027_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 866066 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0