This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03929958_s1
See other 1810017P11RIK GE Assays ›
Gene Symbol:
1810017P11Rik
Gene Name:
RIKEN cDNA 1810017P11 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03929958_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 1810017P11Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK007529.1 - 1 50 96

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream HNF1 homeobox A Hnf1a AI323641,
HNF1,
HNF1-alpha,
HNF1[a],
Hnf-1,
Hnf1alpha,
LFB1,
Tcf-1,
Tcf1
-4.4kb Anti-sense Chr. 5 - 114948980 - 114971062 on NCBI Build GRCm38
Downstream signal peptide peptidase 3 Sppl3 4833416I09Rik,
Psl4,
Usmg3
35.5kb Sense Chr. 5 - 115011524 - 115098790 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Agilent Illumina

Array Name Probe IDs Targets Target Type
Agilent Six_ChIP_01_of_03_v2 A_68_P04889413 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24007188 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P24007186 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04889409 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04889410 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 104670576 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0