This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03929887_s1
See other 1700123E06RIK GE Assays ›
Gene Symbol:
1700123E06Rik
Gene Name:
RIKEN cDNA 1700123E06 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03929887_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 1700123E06Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK007248.1 - 1 652 73

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream tankyrase,
TRF1-interacting ankyrin-related ADP-ribose polymerase
Tnks 4930554K12Rik,
AI662855,
ARTD5,
C86528,
D130072O21Rik,
TANK1,
mTNKS1
-128.8kb Sense Chr. 8 - 34829179 - 34965690 on NCBI Build GRCm38
Downstream protein phosphatase 1,
regulatory (inhibitor) subunit 3B
Ppp1r3b 6430576E21,
AW821953,
GL,
PTG
280.4kb Anti-sense Chr. 8 - 35375668 - 35388139 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

Agilent ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Six_ChIP_02_of_03_v2 A_68_P25777816 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25777823 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25777808 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25777813 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25777824 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25777818 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25777809 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06966875 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06829358 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25777826 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25777827 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06966884 - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 415157 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 106370347 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0