This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03929862_s1
See other TMEM165 GE Assays ›
Gene Symbol:
Tmem165
Gene Name:
transmembrane protein 165
Gene Aliases:
AV026557, Tpardl, Tparl, pFT27
Chromosome Location:
Chr.5: 76183880 - 76209244 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03929862_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tmem165
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK007181.1 - 1 751 59

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream steroid 5 alpha-reductase 3 Srd5a3 1110025P14Rik,
A430076C09,
AV364670,
AW987574,
D730040M03Rik,
H5ar,
S5AR 3,
Srd5a2l
-27.9kb Anti-sense Chr. 5 - 76140273 - 76155503 on NCBI Build GRCm38
Downstream circadian locomotor output cycles kaput Clock 5330400M04Rik,
KAT13D
25.5kb Sense Chr. 5 - 76209868 - 76305007 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0