This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03929853_s1
See other TTLL13 GE Assays ›
Gene Symbol:
Ttll13
Gene Name:
tubulin tyrosine ligase-like family, member 13
Gene Aliases:
1700111A04Rik, 4921538O11, BC024578
Chromosome Location:
Chr.7: 80246376 - 80260821 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03929853_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ttll13
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK007167.1 - 1 257 106
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY707251.1 - 1 257 Chr. 7 - 80246425 - 80246806 [+] on Build GRCm38 106

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream GDP-D-glucose phosphorylase 1 Gdpgp1 D330012F22Rik -5.0kb Sense Chr. 7 - 80232893 - 80241420 on NCBI Build GRCm38
Downstream neugrin,
neurite outgrowth associated
Ngrn AW552001 14.4kb Sense Chr. 7 - 80261215 - 80265378 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0