This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03929717_s1
See other 1700074L02RIK GE Assays ›
Gene Symbol:
1700074L02Rik
Gene Name:
RIKEN cDNA 1700074L02 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03929717_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 1700074L02Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK006950.1 - 1 254 106
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CN666292.1 - 1 413 Chr. 18 - 38298914 - 38299444 [+] on Build GRCm38 106
GenBankmRNA BX629883.1 - 1 Chr. 18 - 38299178 - 38299474 [-] on Build GRCm38 106
GenBankmRNA BY706981.1 - 1 254 Chr. 18 - 38299073 - 38299471 [+] on Build GRCm38 106

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream ring finger protein 14 Rnf14 2310075C09Rik,
2610005D23Rik,
AA986456,
AU041447,
D18Ertd188e,
D7Bwg0165e,
Triad2
Sense Chr. 18 - 38296635 - 38317852 on NCBI Build GRCm38
Downstream glucosamine-6-phosphate deaminase 1 Gnpda1 GNPDA,
Gnp1,
Gnpi,
oscillin
28.1kb Anti-sense Chr. 18 - 38327537 - 38338993 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Illumina

Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 106620427 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0