This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03929706_s1
See other 1700074A11RIK GE Assays ›
Gene Symbol:
1700074A11Rik
Gene Name:
RIKEN cDNA 1700074A11 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03929706_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 1700074A11Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK006945.1 - 1 148 100
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY706969.1 - 1 146 Chr. 2 - 181837417 - 181837798 [-] on Build GRCm38 100

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream protein-L-isoaspartate (D-aspartate) O-methyltransferase domain containing 2 Pcmtd2 5330414D10Rik,
AI314201
Anti-sense Chr. 2 - 181835932 - 181857460 on NCBI Build GRCm38
Downstream polymerase (RNA) III (DNA directed) polypeptide K Polr3k 12.3kDa,
1500004O14Rik,
AI196849,
AU014893,
C11,
RPC10,
RPC11
26.6kb Anti-sense Chr. 2 - 181864360 - 181870830 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0