This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03929700_s1
See other 4931413K12RIK GE Assays ›
Gene Symbol:
4931413K12Rik
Gene Name:
RIKEN cDNA 4931413K12 gene
Gene Aliases:
1700067H02Rik
Chromosome Location:
Chr.11: 75590024 - 75597381 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03929700_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4931413K12Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK006915.1 - 1 249 105
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY706911.1 - 1 246 Chr. 11 - 75594260 - 75594737 [-] on Build GRCm38 105

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream phosphatidylinositol transfer protein,
alpha
Pitpna Pitpn,
vb,
vibrator
Anti-sense Chr. 11 - 75588108 - 75628778 on NCBI Build GRCm38
Downstream cytochrome c oxidase,
subunit VIIc pseudogene
Gm12338 OTTMUSG00000006216 4.9kb Sense Chr. 11 - 75599634 - 75600231 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0