This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03929674_m1
See other 1700060J05RIK GE Assays ›
Gene Symbol:
1700060J05Rik
Gene Name:
RIKEN cDNA 1700060J05 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03929674_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 1700060J05Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK006843.1 - 179 99
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY706824.1 - 1 - 2 179 Chr. 4 - 53563067 - 53564697 [-] on Build GRCm38 99

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream solute carrier family 44,
member 1
Slc44a1 2210409B22Rik,
4833416H08Rik,
AW547365,
CHTL1,
CTL1,
Cdw92
Anti-sense Chr. 4 - 53440413 - 53622478 on NCBI Build GRCm38
Downstream fibronectin type III and SPRY domain containing 1-like Fsd1l A230072O16Rik,
Ccdc10,
Csdufd1,
Fsd1nl,
OTTMUSG00000007002
65.2kb Anti-sense Chr. 4 - 53629929 - 53707009 on NCBI Build GRCm38

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Mus musculus 1700 Probe spans exons

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140380 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1453476_PM_at - -
Mouse Genome 430 2.0 Array 1453476_PM_at - -
Mouse Genome 430 2.0 Array 1453476_at - -
Mouse Expression Set 430 1453476_PM_at - -
Mouse Expression Set 430 1453476_at - -
Affymetrix HT Mouse Genome 430B Array 1453476_at - -
Affymetrix HT Mouse Genome 430B Array 1453476_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 684604 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 100670170 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0