This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03929653_m1
See other 1700055D16RIK GE Assays ›
Gene Symbol:
1700055D16Rik
Gene Name:
RIKEN cDNA 1700055D16 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03929653_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 1700055D16Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK006793.1 - 1 - 2 138 76
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY706775.1 - 1 - 2 134 Chr. 6 - 134792363 - 134820678 [+] on Build GRCm38 76

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream dual specificity phosphatase 16 Dusp16 3830417M17Rik,
AW558566,
D6Ertd213e,
MKP-7,
MKP7,
Mkpm
Anti-sense Chr. 6 - 134715468 - 134792628 on NCBI Build GRCm38
Downstream cAMP responsive element binding protein-like 2 Crebl2 AI046348,
B230205M03
8.0kb Sense Chr. 6 - 134828686 - 134858931 on NCBI Build GRCm38

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Mus musculus 1700 Probe spans exons

MicroArray Probes:

Agilent ABI

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 496290 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 654960 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 362184 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 136364 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 266200 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 151219 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 479197 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 755455 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 283094 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 10487 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 849875 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 817979 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 914858 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 824590 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 338521 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 71558 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 86812 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 61349 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P162171 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P162171 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06076926 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06076934 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P162171 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06076934 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05852225 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24863723 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05625472 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05625477 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05625470 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24863711 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24863724 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24863709 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05625474 - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 408422 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-fd98957d6-rbs6c:80/100.66.128.142:80.
git-commit: 278a916a254c4ca07f9ccbb2b9efa8cd7578b5be
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0