This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03929525_s1
See other 4930518C04RIK GE Assays ›
Gene Symbol:
4930518C04Rik
Gene Name:
RIKEN cDNA 4930518C04 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03929525_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4930518C04Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK006476.1 - 1 350 70
AK006477.1 - 1 351 70
AK015821.1 - 1 334 70
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY706515.1 - 1 351 Chr. 14 - 103899467 - 103899959 [+] on Build GRCm38 70
GenBankmRNA BY715365.1 - 1 334 Chr. 14 - 103899484 - 103900177 [+] on Build GRCm38 70
GenBankmRNA BY706516.1 - 1 351 Chr. 14 - 103899467 - 103899955 [+] on Build GRCm38 70

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream endothelin receptor type B Ednrb ET-B,
ET-BR,
ETR-b,
ETb,
Sox10m1
-55.0kb Anti-sense Chr. 14 - 103814615 - 103844476 on NCBI Build GRCm38
Downstream RIKEN cDNA 4930432J09 gene 4930432J09Rik 77.7kb Anti-sense Chr. 14 - 103977628 - 104027142 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105136 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29868403 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29868404 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29868414 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29868407 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29868408 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29868405 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29868416 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11440219 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29868410 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29868412 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1433259_PM_at - -
Mouse Genome 430 2.0 Array 1454529_at - -
Mouse Genome 430 2.0 Array 1433259_PM_at - -
Mouse Genome 430 2.0 Array 1433259_at - -
Mouse Genome 430 2.0 Array 1454529_PM_at - -
Mouse Expression Set 430 1433259_PM_at - -
Mouse Expression Set 430 1454529_PM_at - -
Mouse Expression Set 430 1454529_at - -
Mouse Expression Set 430 1433259_at - -
Affymetrix HT Mouse Genome 430B Array 1433259_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1433259_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 531958 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 100730348 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0