This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03929396_s1
See other TOPORSL GE Assays ›
Gene Symbol:
Toporsl
Gene Name:
topoisomerase I binding, arginine/serine-rich like
Gene Aliases:
4930547C10Rik
Chromosome Location:
Chr.4: 52596274 - 52612424 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03929396_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Toporsl
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK006061.1 - 1 1953 158
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB679288.1 - 1 111 Chr. 4 - 52612102 - 52612427 [+] on Build GRCm38 158

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream structural maintenance of chromosomes 2 Smc2 5730502P04Rik,
AI255214,
AW545314,
CAP-E,
CAPE,
Fin16,
SMC-2,
Smc2l1
-121.9kb Sense Chr. 4 - 52439221 - 52488365 on NCBI Build GRCm38
Downstream RIKEN cDNA 4930552N02 gene 4930552N02Rik 202.2kb Sense Chr. 4 - 52814604 - 52828269 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7bff9b55f5-ncxmj:80/100.66.128.142:80.
git-commit: 5f36da1a8ad9bc9722227bfd86ebc827ae27d083
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0