This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03929347_s1
See other 1700013M08RIK GE Assays ›
Gene Symbol:
1700013M08Rik
Gene Name:
RIKEN cDNA 1700013M08 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03929347_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 1700013M08Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK005964.1 - 1 652 95

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream Rho GTPase activating protein 24 Arhgap24 0610025G21Rik Anti-sense Chr. 5 - 102207198 - 102897937 on NCBI Build GRCm38
Downstream mitogen-activated protein kinase 10 Mapk10 C230008H04Rik,
JNK3,
JNK3B1,
JNK3B2,
SAPK(beta),
Serk2,
p493F12,
p54bSAPK
428.0kb Sense Chr. 5 - 102907948 - 103211339 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

Agilent ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2345825 AK005964.1 mouse_transcriptome
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 595178 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 101090402 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0