This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03929299_m1
See other D130017N08RIK GE Assays ›
Gene Symbol:
D130017N08Rik
Gene Name:
RIKEN cDNA D130017N08 gene
Gene Aliases:
1700012G11Rik
Chromosome Location:
Chr.5: 143758354 - 143764942 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03929299_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: D130017N08Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK005905.1 - 295 57
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY706045.1 - 1 - 2 295 Chr. 5 - 143759862 - 143767130 [+] on Build GRCm38 57

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream ubiquitin specific peptidase 42 Usp42 2410140K03Rik,
3110031A07Rik,
A630018G05Rik,
D5Ertd591e
-54.0 Anti-sense Chr. 5 - 143710325 - 143759808 on NCBI Build GRCm38
Downstream eukaryotic translation initiation factor 2 alpha kinase 1 Eif2ak1 HCR,
Hri
108.1kb Sense Chr. 5 - 143871862 - 143902717 on NCBI Build GRCm38

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mus musculus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0