This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03929248_s1
See other 1700008F19RIK GE Assays ›
Gene Symbol:
1700008F19Rik
Gene Name:
RIKEN cDNA 1700008F19 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03929248_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 1700008F19Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK005763.1 - 1 847 89
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BQ032111.1 - 1 Chr. 19 - 41264815 - 41265041 [-] on Build GRCm38 89
GenBankmRNA BX633045.1 - 1 Chr. 19 - 41264765 - 41265036 [-] on Build GRCm38 89

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream transmembrane 9 superfamily member 3 Tm9sf3 1810073M23Rik,
2810031D16Rik,
AI115521,
AI413748,
AW146116,
AW549777,
Smbp,
mKIAA4036
-108.0 Anti-sense Chr. 19 - 41210842 - 41264004 on NCBI Build GRCm38
Downstream phosphoinositide-3-kinase adaptor protein 1 Pik3ap1 1810044J04Rik,
BCAP
7.7kb Anti-sense Chr. 19 - 41272752 - 41385070 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130782 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1441822_PM_at - -
Mouse Genome 430 2.0 Array 1441822_PM_at - -
Mouse Genome 430 2.0 Array 1441822_at - -
Mouse Expression Set 430 1441822_PM_at - -
Mouse Expression Set 430 1441822_at - -
Affymetrix HT Mouse Genome 430B Array 1441822_at - -
Affymetrix HT Mouse Genome 430B Array 1441822_PM_at - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 103850672 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0