Assay Details
Gene Symbol: 1700008A23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK005751.1 | - | 1 | 455 | 88 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | extended synaptotagmin-like protein 3 | Esyt3 |
D930024E11, D9Ertd280e, Fam62c, mKIAA4186 |
Anti-sense | Chr. 9 - 99308739 - 99360405 on NCBI Build GRCm38 | |
Downstream | muscle and microspikes RAS | Mras |
2900078C09Rik, AI326250 |
61.9kb | Anti-sense | Chr. 9 - 99385420 - 99436961 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
