This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03929215_m1
See other 1700007J10RIK GE Assays ›
Gene Symbol:
1700007J10Rik
Gene Name:
RIKEN cDNA 1700007J10 gene
Gene Aliases:
4930573L17Rik
Chromosome Location:
Chr.11: 59725917 - 59740154 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03929215_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 1700007J10Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_045476.1 1 - 2 121 99
GenBank mRNA AK005722.1 - 1 - 2 123 99

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream olfactory receptor 225 Olfr225 MOR256-51 -121.7kb Anti-sense Chr. 11 - 59610786 - 59614225 on NCBI Build GRCm38
Downstream phospholipase D family,
member 6
Pld6 4933433K01Rik,
Gm10,
Zuc,
mZuc,
mitoPLD
43.7kb Sense Chr. 11 - 59783893 - 59787657 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100002858.mm.1

Set Membership:

Amplicon length between 86 and 100 Mus musculus Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100025924.mm.1 NR_045476.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100025924.mm.1 NR_045476.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100025924.mm.1 TC1100002858.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100025925.mm.1 NR_045476.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100025925.mm.1 NR_045476.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100025925.mm.1 TC1100002858.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0