This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03929161_s1
See other 1600017P15RIK GE Assays ›
Gene Symbol:
1600017P15Rik
Gene Name:
RIKEN cDNA 1600017P15 gene
Gene Aliases:
Gm24413
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03929161_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 1600017P15Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK005488.1 - 1 152 111

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream MDS1 and EVI1 complex locus Mecom D630039M04Rik,
Evi-1,
Evi1,
Jbo,
Mds,
Mds1,
Mds1-Evi1,
Prdm3,
Znfpr1b1
-85.1kb Sense Chr. 3 - 29951296 - 30509783 on NCBI Build GRCm38
Downstream actin related protein T3 Actrt3 1700119I24Rik,
ARP-T3,
Arpm1
1.3kb Sense Chr. 3 - 30597073 - 30599870 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix ABI

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 932543 - -
Whole Mouse Genome G4122A A_52_P914857 AK005488.1 mouse_transcriptome
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10497616 - -
Affymetrix Mouse Gene 1.1 ST Array 10497615 - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 692607 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0