This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03929159_s1
See other 1600015H23RIK GE Assays ›
Gene Symbol:
1600015H23Rik
Gene Name:
RIKEN cDNA 1600015H23 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03929159_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 1600015H23Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK005466.1 - 1 254 83
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY705668.1 - 1 254 Chr. 11 - 109344286 - 109344633 [+] on Build GRCm38 83

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream regulator of G-protein signaling 9 Rgs9 RGS9-1,
Rgs9-2
-46.1kb Anti-sense Chr. 11 - 109225348 - 109298181 on NCBI Build GRCm38
Downstream RIKEN cDNA 1700096J18 gene 1700096J18Rik 2.2kb Sense Chr. 11 - 109346867 - 109353651 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

Agilent ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Six_ChIP_02_of_03_v2 A_68_P28118047 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09292916 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28118003 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09292932 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28118014 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28117995 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28118000 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28118026 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28118017 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28118046 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09608212 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28118044 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28118024 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28117997 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28118001 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28118042 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28118032 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28118019 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09608211 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28118015 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28117998 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28118022 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28118021 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28118034 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28118013 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09239516 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28117992 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28118031 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28118029 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09466847 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28117994 - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 822202 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 103170397 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0