This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03682602_m1
See other BACH2OS GE Assays ›
Gene Symbol:
Bach2os
Gene Name:
BTB and CNC homology 2, opposite strand
Gene Aliases:
-
Chromosome Location:
Chr.4: 32559680 - 32571662 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03682602_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Bach2os
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_026843.1 2 - 3 127 74
GenBank mRNA BC024582.1 - 2 - 3 127 74
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BI150671.1 - 2 - 3 128 Chr. 4 - 32561587 - 32571662 [-] on Build GRCm38 74

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream mitogen-activated protein kinase kinase kinase 7 Map3k7 B430101B05,
C87327,
Tak1
-539.6kb Anti-sense Chr. 4 - 31962296 - 32023470 on NCBI Build GRCm38
Downstream gap junction protein,
alpha 10
Gja10 Cx-57,
Cx59,
cx57
35.3kb Sense Chr. 4 - 32600865 - 32602382 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400002362.mm.1

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400019188.mm.1 NR_026843.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400019188.mm.1 NR_026843.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400019188.mm.1 TC0400002362.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400019186.mm.1 NR_026843.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400019186.mm.1 NR_026843.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400019186.mm.1 TC0400002362.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400019190.mm.1 NR_026843.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400019190.mm.1 NR_026843.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400019190.mm.1 TC0400002362.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400019191.mm.1 NR_026843.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400019191.mm.1 NR_026843.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400019191.mm.1 TC0400002362.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0