This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03652642_s1
See other NPHS1OS GE Assays ›
Gene Symbol:
Nphs1os
Gene Name:
nephrosis 1 homolog, nephrin, opposite strand
Gene Aliases:
Nphs1as
Chromosome Location:
Chr.7: 30462164 - 30465591 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03652642_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nphs1os
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_004443.1 3 1049 78
GenBank mRNA AY422730.1 - 3 1049 78
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BE979346.1 - 1 Chr. 7 - 30462164 - 30462622 [+] on Build GRCm38 78

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream kin of IRRE like 2 (Drosophila) Kirrel2 C330019F22Rik,
NEPH3,
NLG1
-1.5kb Sense Chr. 7 - 30447530 - 30460680 on NCBI Build GRCm38
Downstream proline dehydrogenase (oxidase) 2 Prodh2 2510028N04Rik,
2510038B11Rik,
MmPOX,
MmPOX1,
POX1
30.6kb Anti-sense Chr. 7 - 30493586 - 30513402 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Agilent Affymetrix

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2004576 NR_004443.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2004576 AY422730.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2004576 NR_004443.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2004576 AY422730.1 mouse_transcriptome
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1447291_PM_at - -
Mouse Genome 430 2.0 Array 1447291_at - -
Mouse Genome 430 2.0 Array 1447291_PM_at - -
Affymetrix Mouse Clariom S Array PSR0700024512.mm.1 NR_004443.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700024512.mm.1 TC0700002761.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700024510.mm.1 NR_004443.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700024510.mm.1 TC0700002761.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700024511.mm.1 NR_004443.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700024511.mm.1 TC0700002761.mm.1 MTA TC
Mouse Expression Set 430 1447291_PM_at - -
Mouse Expression Set 430 1447291_at - -
Affymetrix HT Mouse Genome 430B Array 1447291_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1447291_at - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0