This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03456613_s1
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Species:
Mouse
Assay Type:
Non-coding
Assay ID Mm03456613_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AJ249895.1 - Chr. - - [-] on Build 82
GenBankmRNA BQ944383.1 - 249 Chr. - - [-] on Build 82
GenBankmRNA AI464015.1 - 1 349 Chr. 17 - 12754279 - 12754683 [+] on Build GRCm38 82
GenBankmRNA AA855681.1 - 1 348 Chr. 17 - 12754280 - 12754680 [+] on Build GRCm38 82

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream solute carrier family 22 (organic cation transporter),
member 1
Slc22a1 Lx1,
Oct1,
Orct,
Orct1
-78.4kb Anti-sense Chr. 17 - 12648874 - 12675838 on NCBI Build GRCm38
Downstream MAS1 oncogene Mas1 Mas-1,
Mgra
84.2kb Anti-sense Chr. 17 - 12838841 - 12868145 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0