This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03456609_s1
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Species:
Mouse
Assay Type:
Non-coding
Assay ID Mm03456609_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AJ249895.1 - Chr. - - [-] on Build 90
GenBankmRNA AA756700.1 - 1 44 Chr. 17 - 12753012 - 12753478 [+] on Build GRCm38 90
GenBankmRNA BX523972.1 - 1 44 Chr. 17 - 12753012 - 12753537 [+] on Build GRCm38 90
GenBankmRNA AI425659.1 - 1 44 Chr. 17 - 12753012 - 12753405 [+] on Build GRCm38 90
GenBankmRNA BE631408.1 - 1 44 Chr. 17 - 12753012 - 12753504 [+] on Build GRCm38 90
GenBankmRNA CV675693.1 - 1 294 Chr. 17 - 12752777 - 12753664 [+] on Build GRCm38 90
GenBankmRNA DT915167.1 - 1 247 Chr. 17 - 12752810 - 12753367 [+] on Build GRCm38 90
GenBankmRNA CA947506.1 - 1 334 Chr. 17 - 12752777 - 12753199 [+] on Build GRCm38 90
GenBankmRNA BU581823.1 - 1 312 Chr. 17 - 12752777 - 12753183 [+] on Build GRCm38 90
GenBankmRNA BI793861.1 - 1 287 Chr. 17 - 12752777 - 12753258 [+] on Build GRCm38 90

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream solute carrier family 22 (organic cation transporter),
member 1
Slc22a1 Lx1,
Oct1,
Orct,
Orct1
-77.2kb Anti-sense Chr. 17 - 12648874 - 12675838 on NCBI Build GRCm38
Downstream MAS1 oncogene Mas1 Mas-1,
Mgra
85.4kb Anti-sense Chr. 17 - 12838841 - 12868145 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0