This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03456590_s1
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Species:
Mouse
Assay Type:
Non-coding
Assay ID Mm03456590_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AJ249895.1 - Chr. - - [-] on Build 107
GenBankmRNA AA175448.1 - 1 243 Chr. 17 - 12772258 - 12772692 [+] on Build GRCm38 107
GenBankmRNA AK138959.1 - 934 Chr. - - [-] on Build 107
GenBankmRNA AI645109.1 - 1 243 Chr. 17 - 12772258 - 12772738 [+] on Build GRCm38 107
GenBankmRNA BQ554196.1 - 1 484 Chr. 17 - 12772020 - 12772655 [+] on Build GRCm38 107
GenBankmRNA BF452763.1 - 1 243 Chr. 17 - 12772258 - 12772641 [+] on Build GRCm38 107

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream insulin-like growth factor 2 receptor Igf2r AI661837,
CD222,
CI-MPR,
M6P/IGF2R,
Mpr300
-2.6kb Anti-sense Chr. 17 - 12682406 - 12769706 on NCBI Build GRCm38
Downstream MAS1 oncogene Mas1 Mas-1,
Mgra
66.1kb Anti-sense Chr. 17 - 12838841 - 12868145 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0