This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03455923_s1
See other D630033A02RIK GE Assays ›
Gene Symbol:
D630033A02Rik Mirlet7i
Gene Name:
RIKEN cDNA D630033A02 gene
microRNA let7i
Gene Aliases:
-
Mirnlet7i, let-7i, mmu-let-7i
Chromosome Location:
-
Chromosome Location:
Chr.10: 122985640 - 122985724 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
Assay ID Mm03455923_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: D630033A02Rik
Interrogated Sequence Translated Protein RNADB ID Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK052706.1 - LIT3440
D630033A02Rik
153 70
Gene Symbol: Mirlet7i
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_029527.1 1 45 70
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB661268.1 - 1 153 Chr. 10 - 122985221 - 122985831 [-] on Build GRCm38 70
GenBankmRNA CJ179156.1 - 1 116 Chr. 10 - 122985399 - 122985794 [-] on Build GRCm38 70

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream protein phosphatase 1H (PP2C domain containing) Ppm1h A430075L18Rik,
ARHCL1,
C030002B11Rik
-39.6kb Anti-sense Chr. 10 - 122678106 - 122945795 on NCBI Build GRCm38
Downstream MON2 homolog (yeast) Mon2 2610528O22Rik,
AW495628,
Sf21,
mKIAA1040
6.3kb Sense Chr. 10 - 122992060 - 123076505 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s)

cellular response to amino acid stimulus

MicroArray Probes:

Agilent Affymetrix ABI

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2063937 BE691013.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 9581 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27511717 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27511719 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27511720 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1447925_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10372926 - -
Mouse Genome 430 2.0 Array 1447925_PM_at - -
Mouse Genome 430 2.0 Array 1447925_at - -
Affymetrix Mouse Clariom S Array PSR1000022844.mm.1 NR_029527.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000022844.mm.1 NR_029527.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000022844.mm.1 TC1000003065.mm.1 MTA TC
Mouse Expression Set 430 1447925_PM_at - -
Mouse Expression Set 430 1447925_at - -
Affymetrix HT Mouse Genome 430B Array 1447925_at - -
Affymetrix HT Mouse Genome 430B Array 1447925_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 464022 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0