This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03455590_m1
See other IGF2OS GE Assays ›
Gene Symbol:
Igf2os
Gene Name:
insulin-like growth factor 2, opposite strand
Gene Aliases:
Igf2as, Peg8
Chromosome Location:
Chr.7: 142659693 - 142670356 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03455590_m1
Size
Availability Made To Order
Catalog # 4426961
Price 4,437.00
Your Price
Online offer:
4437.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Igf2os
Interrogated Sequence Translated Protein RNADB ID Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_002855.2
2 - 3 1396 84
GenBank mRNA AB030734.1 - LIT1673
PEG8/Igf2as
2 - 3 1395 84

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream insulin-like growth factor 2 Igf2 AL033362,
Igf-2,
Igf-II,
M6pr,
Mpr,
Peg2
-3.3kb Anti-sense Chr. 7 - 142650768 - 142662270 on NCBI Build GRCm38
Downstream insulin II Ins2 AA986540,
Ins-2,
InsII,
Mody,
Mody4,
proinsulin
12.2kb Anti-sense Chr. 7 - 142678656 - 142679708 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700002084.mm.1

Set Membership:

Amplicon length between 71 and 85 Mus musculus Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700018776.mm.1 NR_002855.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700018776.mm.1 TC0700002084.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0