This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03053778_s1
See other NFATC2IP GE Assays ›
Gene Symbol:
Nfatc2ip
Gene Name:
nuclear factor of activated T cells, cytoplasmic, calcineurin dependent 2 interacting protein
Gene Aliases:
D7Ertd304e, NIP45
Chromosome Location:
Chr.7: 126382853 - 126397252 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03053778_s1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nfatc2ip
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010900.3 NP_035030.2 8 3203 126
GenBank mRNA AK030107.1 - 8 3205 126

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Inventoried Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Nfatc2ip

Gene Ontology Categories:

Function(s) Process(es)

cytokine production
positive regulation of transcription from RNA polymerase II promoter
protein binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25500259 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25500261 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25500262 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25500258 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25500257 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25500260 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P646034 AK030107.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P646034 NM_010900.3 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P646034 AK030107.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P646034 NM_010900.3 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 342400 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 769791 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 437309 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 9463 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 517325 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 928211 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 55250 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 44448 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P104351 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P167811 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P105951 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P646034 AK030107.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P646034 NM_010900.3 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P646034 AK030107.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P646034 NM_010900.3 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P201337 BC113761.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P201337 U76759.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P201337 AK030107.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P201337 NM_010900.3 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P201337 AK005947.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500265 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500285 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500256 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500271 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500250 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500272 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500267 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500287 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500195 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500259 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500278 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500258 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500254 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500276 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500264 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500270 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500262 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500251 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500266 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500273 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500248 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500253 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500246 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500260 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500269 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500284 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25500257 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06358776 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P105951 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P104351 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01629657 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P167811 - -
Whole Mouse Genome G4122A A_52_P646034 AK030107.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P646034 NM_010900.3 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P646034 AK030107.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P646034 NM_010900.3 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P201337 BC113761.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P201337 U76759.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P201337 AK030107.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P201337 NM_010900.3 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P201337 AK005947.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100367 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P119046 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P167811 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P104351 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P105951 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06119673 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06119680 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06119677 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06119676 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25500258 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25500257 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25500260 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25500259 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25500261 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25500262 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500155 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500206 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500256 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500254 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500215 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500285 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500240 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500216 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500230 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500146 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500241 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500222 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500233 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500176 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500208 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500182 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500301 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500272 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500203 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500195 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500199 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500278 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500194 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500218 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500212 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500149 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500185 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500253 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500189 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500286 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500138 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500159 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500137 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500133 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500177 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500140 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500248 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500143 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500131 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500172 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500236 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500191 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500187 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500200 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500135 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500260 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500165 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500271 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500250 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500255 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500304 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500153 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500292 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500150 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500262 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500168 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500217 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500225 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500277 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500259 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500303 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500246 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500231 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500167 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500211 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500145 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500152 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500220 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500299 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500269 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500174 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500193 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500251 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500237 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500234 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500163 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500161 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500192 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500213 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500169 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500221 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500294 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500265 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500291 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500197 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25500179 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06034122 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25500245 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25500271 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25500278 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25500296 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25500280 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25500221 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05994205 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25500237 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25500227 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25500212 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25500236 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06398838 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25500244 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25500222 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25500254 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25500264 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25500229 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25500255 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06264226 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25500200 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25500203 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25500292 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25500206 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25500230 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25500273 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25500241 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25500211 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25500248 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25500197 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25500284 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06314876 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25500256 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25500246 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25500240 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25500257 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25500231 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25500253 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25500243 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25500195 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06119677 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25500199 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25500262 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25500270 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25500294 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25500225 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06053842 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25500242 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25500285 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06446655 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25500250 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25500272 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25500266 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25500228 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25500208 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25500234 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25500233 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06543878 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06119676 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25500198 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25500287 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25500265 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25500291 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06534596 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10567852 - -
Affymetrix Mouse Gene 1.0 ST Array 10567859 - -
Affymetrix Mouse Gene 1.0 ST Array 10567858 - -
Affymetrix Mouse Gene 1.0 ST Array 10567860 - -
Affymetrix Mouse Gene 1.0 ST Array 10567862 - -
Affymetrix Mouse Gene 1.0 ST Array 10567856 - -
Affymetrix Mouse Gene 1.0 ST Array 10567857 - -
Affymetrix Mouse Gene 1.0 ST Array 10567861 - -
Affymetrix Mouse Gene 1.0 ST Array 10567854 - -
Affymetrix Mouse Gene 1.0 ST Array 10567853 - -
Affymetrix Mouse Gene 1.0 ST Array 10567855 - -
Affymetrix HT MG-430 PM Array Plate 1434836_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1451781_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10567851 - -
Mouse Genome 430 2.0 Array 1451781_PM_at - -
Mouse Genome 430 2.0 Array 1451781_at - -
Mouse Genome 430 2.0 Array 1434836_PM_at - -
Mouse Genome 430 2.0 Array 1434836_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 18020_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 18020_PM_at - -
Affymetrix Mouse Clariom S Array PSR0700035620.mm.1 NM_010900.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700035620.mm.1 NM_010900.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700035620.mm.1 TC0700004276.mm.1 MTA TC
Mouse Expression Set 430 1434836_at - -
Mouse Expression Set 430 1434836_PM_at - -
Mouse Expression Set 430 1451781_at - -
Mouse Expression Set 430 1451781_PM_at - -
Murine Genome U74v2 Set 100699_PM_at - -
Murine Genome U74v2 Set 96561_PM_at - -
Murine Genome U74v2 Set 100699_at - -
Murine Genome U74v2 Set 96561_at - -
Mouse Genome 430A 2.0 Array 1434836_PM_at - -
Mouse Genome 430A 2.0 Array 1451781_PM_at - -
Mouse Genome 430A 2.0 Array 1451781_at - -
Mouse Genome 430A 2.0 Array 1434836_at - -
Affymetrix HT Mouse Genome 430A Array 1434836_at - -
Affymetrix HT Mouse Genome 430A Array 1434836_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1451781_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1451781_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 405951 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 5130687 - -
Illumina mouse-6 v1.0 expression beadchip 5130687 - -
Illumina mouse-6 v1.0 expression beadchip 106400133 - -
Illumina mouseRef-8 v1.0 expression beadchip 0005130687 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1235902 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1243004 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1235902 - -
mouse-6 v1.1 (Illumina) scl30690.8_420-S - -
mouse-6 v1.1 (Illumina) scl9310.1.1_141-S - -
Illumina Sentrix mouse-6.v1 bead array 5130687 - -
Illumina Sentrix mouse-6.v1 bead array 106400133 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl30690.8_420-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 5130687 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 106400133 - -
MouseRef-8 v2 Expression BeadChip ILMN_1235902 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1235902 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl30690.8_420-S - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0