This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03049626_s1
See other TSPYL1 GE Assays ›
Gene Symbol:
Tspyl1
Gene Name:
testis-specific protein, Y-encoded-like 1
Gene Aliases:
Tspyl
Chromosome Location:
Chr.10: 34282190 - 34284885 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03049626_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tspyl1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009433.3 NP_033459.1 1 205 130
GenBank mRNA AF042180.1 - 1 169 130
AK053812.1 - 1 156 130
AK159432.1 - 1 186 130
AK166438.1 - 1 184 130
AK166805.1 - 1 134 130
AK166893.1 - 1 134 130
BC011213.1 - 1 152 4196841 130
BY281503.1 - 1 205 130

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tspyl1

Panther Classification:

Molecular Function -

chromatin/chromatin-binding, or -regulatory protein

Gene Ontology Categories:

Function(s) Process(es)

nucleosome assembly
enzyme binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000001981.mm.1 NM_009433.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000001981.mm.1 NM_009433.3 Refseq
Affymetrix Mouse Clariom S Array PSR1000001981.mm.1 TC1000000297.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000001982.mm.1 NM_009433.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000001982.mm.1 NM_009433.3 Refseq
Affymetrix Mouse Clariom S Array PSR1000001982.mm.1 TC1000000297.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 681498 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0