This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03038599_m1
See other PCDH9 GE Assays ›
Gene Symbol:
Pcdh9
Gene Name:
protocadherin 9
Gene Aliases:
-
Chromosome Location:
Chr.14: 93013700 - 93890721 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03038599_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pcdh9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081377.2 NP_001074846.1 1 - 2 3191 104
GenBank mRNA AY861423.1 - 1 - 2 527 104
BC158117.1 - 1 - 2 3111 9088190 104

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400002534.mm.1

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

cadherin

Gene Ontology Categories:

Function(s) Process(es)

biological_process
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400019346.mm.1 NM_001081377.2 Refseq
Affymetrix Mouse Clariom S Array PSR1400019346.mm.1 NM_001271798.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400019346.mm.1 NM_001271799.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400019346.mm.1 NM_001271800.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400019346.mm.1 TC1400002534.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400019347.mm.1 NM_001081377.2 Refseq
Affymetrix Mouse Clariom S Array PSR1400019347.mm.1 NM_001271798.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400019347.mm.1 NM_001271800.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400019347.mm.1 TC1400002534.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400019349.mm.1 NM_001081377.2 Refseq
Affymetrix Mouse Clariom S Array PSR1400019349.mm.1 NM_001271798.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400019349.mm.1 NM_001271800.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400019349.mm.1 TC1400002534.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400019345.mm.1 NM_001081377.2 Refseq
Affymetrix Mouse Clariom S Array PSR1400019345.mm.1 TC1400002534.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400019351.mm.1 NM_001081377.2 Refseq
Affymetrix Mouse Clariom S Array PSR1400019351.mm.1 NM_001271798.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400019351.mm.1 NM_001271800.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400019351.mm.1 TC1400002534.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0