This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03024063_m1
See other TGFBR3 GE Assays ›
Gene Symbol:
Tgfbr3
Gene Name:
transforming growth factor, beta receptor III
Gene Aliases:
1110036H20Rik, AU015626, AW215636, TBRIII
Chromosome Location:
Chr.5: 107106570 - 107289595 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03024063_m1
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tgfbr3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011578.3 NP_035708.2 8 - 9 1512 144
GenBank mRNA AF039601.1 - 8 - 9 1312 144
AK133234.1 - 6 - 7 1022 144
BC070428.1 - 8 - 9 1437 6410554 144

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tgfbr3

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

blood vessel development
vasculogenesis
osteoblast differentiation
in utero embryonic development
blastocyst development
epithelial to mesenchymal transition
liver development
blood vessel remodeling
epicardial cell to mesenchymal cell transition
protein complex assembly
protein phosphorylation
immune response
transforming growth factor beta receptor signaling pathway
transforming growth factor beta receptor complex assembly
positive regulation of cell proliferation
embryo development
negative regulation of epithelial cell migration
negative regulation of epithelial to mesenchymal transition
cell growth
BMP signaling pathway
positive regulation of transforming growth factor beta receptor signaling pathway
negative regulation of transforming growth factor beta receptor signaling pathway
positive regulation of BMP signaling pathway
response to follicle-stimulating hormone
response to prostaglandin E
response to luteinizing hormone
intracellular signal transduction
regulation of protein binding
negative regulation of epithelial cell proliferation
regulation of blood vessel size
positive regulation of NF-kappaB transcription factor activity
negative regulation of cellular component movement
ventricular cardiac muscle tissue morphogenesis
palate development
cardiac muscle cell proliferation
definitive hemopoiesis
cardiac epithelial to mesenchymal transition
definitive erythrocyte differentiation
heart trabecula formation
pathway-restricted SMAD protein phosphorylation
coronary vasculature development
coronary vasculature morphogenesis
vasculogenesis involved in coronary vascular morphogenesis
visceral serous pericardium development
negative regulation of apoptotic process involved in morphogenesis
transforming growth factor beta-activated receptor activity
type II transforming growth factor beta receptor binding
transforming growth factor beta receptor binding
protein binding
glycosaminoglycan binding
heparin binding
coreceptor activity
fibroblast growth factor binding
PDZ domain binding
SMAD binding
activin binding
transforming growth factor beta binding
transforming growth factor beta receptor activity, type III

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0