This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03011545_g1
See other TMX2 GE Assays ›
Gene Symbol:
Tmx2
Gene Name:
thioredoxin-related transmembrane protein 2
Gene Aliases:
2310042M24Rik, AA589631, Txndc14
Chromosome Location:
Chr.2: 84671311 - 84678828 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03011545_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tmx2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001290751.1 NP_001277680.1 6 - 7 764 96
NM_025868.4 NP_080144.1 8 - 9 1064 96
GenBank mRNA AK009759.1 - 7 - 8 752 96
AK145245.1 - 7 - 8 773 96
AK170587.1 - 7 - 8 761 96
BC013544.1 - 7 - 8 746 4242418 96
BC110994.1 - 7 - 8 747 5349078 96

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tmx2

Panther Classification:

Molecular Function -

oxidoreductase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

biological_process
cell redox homeostasis
molecular_function

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2023777 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2052495 BC110994.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2052495 AK145245.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2052495 AK170587.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2052495 BC013544.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2052495 NM_025868.3 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 566930 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 235285 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 472722 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 104287 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 79853 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2052495 BC110994.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2052495 AK145245.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2052495 AK170587.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2052495 BC013544.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2052495 NM_025868.3 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2023777 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21393950 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21393965 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21393956 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21393955 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21393954 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21393963 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21393971 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21393949 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21393964 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21393974 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21393940 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21393957 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21393969 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21393979 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21393945 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21393970 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21393967 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21393978 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21393972 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21393959 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21393973 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21393942 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21393947 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04591122 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04591140 - -
Whole Mouse Genome G4122A A_52_P617080 BC110994.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P617080 AK145245.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P617080 AK170587.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P617080 BC013544.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P617080 NM_025868.3 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P537492 BC110994.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P537492 AK145245.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P537492 AK170587.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P537492 BC013544.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P537492 NM_025868.3 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P135477 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P04591122 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01279412 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01279406 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01279397 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01279417 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01279404 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21393970 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21393978 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21393925 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21393965 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21393915 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21393888 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21393942 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21393955 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21393957 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21393930 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21393963 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21393947 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21393918 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21393954 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21393949 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21393945 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21393967 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21393940 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21393912 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21393964 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21393921 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21393969 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21393882 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21393979 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21393916 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21393928 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21393973 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21393972 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21393923 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21393971 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21393956 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21393959 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21393970 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21393959 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21393954 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21393942 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21393939 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21393956 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21393972 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21393971 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21393969 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21393967 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21393945 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01696180 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01952926 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21393940 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21393950 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21393955 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21393957 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21393963 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01279413 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21393947 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10484438 - -
Affymetrix Mouse Gene 1.0 ST Array 10484435 - -
Affymetrix Mouse Gene 1.0 ST Array 10484437 - -
Affymetrix Mouse Gene 1.0 ST Array 10484436 - -
Affymetrix Mouse Gene 1.0 ST Array 10484433 - -
Affymetrix Mouse Gene 1.0 ST Array 10484439 - -
Affymetrix Mouse Gene 1.0 ST Array 10484432 - -
Affymetrix Mouse Gene 1.0 ST Array 10484434 - -
Affymetrix Mouse Gene 1.0 ST Array 10447355 - -
Affymetrix HT MG-430 PM Array Plate 1437454_PM_a_at - -
Affymetrix HT MG-430 PM Array Plate 1452130_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10447354 - -
Affymetrix Mouse Gene 1.1 ST Array 10484431 - -
Mouse Genome 430 2.0 Array 1452130_PM_at - -
Mouse Genome 430 2.0 Array 1437454_a_at - -
Mouse Genome 430 2.0 Array 1452130_at - -
Mouse Genome 430 2.0 Array 1437454_PM_a_at - -
Affymetrix Mouse Clariom S Array PSR0200033507.mm.1 NM_025868.4 Refseq
Affymetrix Mouse Clariom S Array PSR0200033507.mm.1 TC0200003873.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200033506.mm.1 NM_025868.4 Refseq
Affymetrix Mouse Clariom S Array PSR0200033506.mm.1 TC0200003873.mm.1 MTA TC
Mouse Expression Set 430 1452130_at - -
Mouse Expression Set 430 1437454_a_at - -
Mouse Expression Set 430 1437454_PM_a_at - -
Mouse Expression Set 430 1452130_PM_at - -
Murine Genome U74v2 Set 96649_at - -
Murine Genome U74v2 Set 96649_PM_at - -
Mouse Genome 430A 2.0 Array 1437454_a_at - -
Mouse Genome 430A 2.0 Array 1452130_at - -
Mouse Genome 430A 2.0 Array 1437454_PM_a_at - -
Mouse Genome 430A 2.0 Array 1452130_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 321856 - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.0 expression beadchip 6840286 - -
Illumina mouse-6 v1.0 expression beadchip 6400725 - -
Illumina mouseRef-8 v1.0 expression beadchip 0006840286 - -
Illumina mouseRef-8 v1.0 expression beadchip 0006400725 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2755355 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2598532 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2755355 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2598532 - -
mouse-6 v1.1 (Illumina) scl0003075.1_15-S - -
mouse-6 v1.1 (Illumina) scl066958.2_91-S - -
Illumina Sentrix mouse-6.v1 bead array 6400725 - -
Illumina Sentrix mouse-6.v1 bead array 6840286 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl066958.2_91-S - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl0003075.1_15-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 6840286 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 6400725 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2598532 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2755355 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl066958.2_91-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0003075.1_15-S - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-6d4c5dbbff-7nlnn:80/100.66.128.142:80.
git-commit: 3992c1c7134a3a3ff95f78ba9fe94218727d3638
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0