This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03003532_u1
See other B2M GE Assays ›
Gene Symbol:
B2m
Gene Name:
beta-2 microglobulin
Gene Aliases:
Ly-m11, beta2-m, beta2m
Chromosome Location:
Chr.2: 122147687 - 122153082 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03003532_u1
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: B2m
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009735.3 NP_033865.2 2 160 247
GenBank mRNA AK019389.1 - 2 163 247
AK020632.1 - 2 162 247
AK028378.1 - 2 162 247
AK153255.1 - 2 162 247
AK166067.1 - 2 163 247
BC085164.1 - 2 115 6846389 247
X01838.1 - 2 161 247

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Inventoried Amplicon spans exons and probe does not span exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene B2m

Panther Classification:

Molecular Function -

major histocompatibility complex protein

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of T cell mediated cytotoxicity
response to molecule of bacterial origin
immune system process
antigen processing and presentation of peptide antigen via MHC class I
antigen processing and presentation of exogenous protein antigen via MHC class Ib, TAP-dependent
cellular defense response
T cell differentiation in thymus
protein refolding
iron ion homeostasis
negative regulation of receptor binding
glycoprotein binding
protein binding
identical protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0