This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm02024198_s1
See other 4933427I18RIK GE Assays ›
Gene Symbol:
4933427I18Rik
Gene Name:
RIKEN cDNA 4933427I18 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm02024198_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4933427I18Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK016954.1 - 3 956 96

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream secretagogin,
EF-hand calcium binding protein
Scgn -75.6kb Anti-sense Chr. 13 - 23953456 - 23991337 on NCBI Build GRCm38
Downstream cytidine monophospho-N-acetylneuraminic acid hydroxylase Cmah 259.7kb Sense Chr. 13 - 24326980 - 24477289 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the AK016954.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P652013 AK016954.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P652013 AK016954.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P652013 AK016954.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P652013 AK016954.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126214 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10404131 - -
Affymetrix Mouse Gene 1.0 ST Array 10404130 - -
Affymetrix Mouse Gene 1.1 ST Array 10404129 - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 507097 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 102230735 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0