This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm02023920_s1
See other 1700085D07RIK GE Assays ›
Gene Symbol:
1700085D07Rik
Gene Name:
RIKEN cDNA 1700085D07 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm02023920_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 1700085D07Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK007005.1 - 2 258 60
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY707061.1 - 2 254 Chr. 15 - 73249546 - 73264120 [+] on Build GRCm38 60
GenBankmRNA AV206213.1 - 1 221 Chr. 15 - 73263860 - 73264120 [+] on Build GRCm38 60

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream PTK2 protein tyrosine kinase 2 Ptk2 FAK,
FRNK,
Fadk,
mKIAA4203,
p125FAK
Anti-sense Chr. 15 - 73205102 - 73423275 on NCBI Build GRCm38
Downstream DENN/MADD domain containing 3 Dennd3 AI447457,
E030003N15Rik
248.4kb Sense Chr. 15 - 73512560 - 73572242 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 797429 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0