This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm02020171_s1
See other 4633402D09RIK GE Assays ›
Gene Symbol:
4633402D09Rik
Gene Name:
RIKEN cDNA 4633402D09 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm02020171_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4633402D09Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK014620.1 - 2 1091 135

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream gephyrin Gphn 5730552E08Rik,
AI662856,
BC027112,
C230040D23,
GPH,
GPHRYN,
geph
Anti-sense Chr. 12 - 78224153 - 78684772 on NCBI Build GRCm38
Downstream family with sequence similarity 71,
member D
Fam71d 4921509E07Rik,
4930516C23Rik,
GARI-L2
328.3kb Anti-sense Chr. 12 - 78691488 - 78734519 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2228953 AK014620.1 mouse_transcriptome
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1432247_PM_at - -
Mouse Genome 430 2.0 Array 1432247_PM_at - -
Mouse Genome 430 2.0 Array 1432247_at - -
Mouse Expression Set 430 1432247_at - -
Mouse Expression Set 430 1432247_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1432247_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1432247_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 890207 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 102320044 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0